Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111077.1 | NP_001104547.1 | 8 | 904 | ||
NM_003379.4 | NP_003370.2 | 7 | 938 | |||
XM_011536110.1 | XP_011534412.1 | 4 | 315 | |||
GenBank mRNA | AF187552.1 | - | 2 | 151 | ||
AF188897.1 | - | 2 | 133 | |||
AF189213.1 | - | 2 | 133 | |||
AF190059.1 | - | 2 | 125 | |||
AF199015.1 | - | 2 | 135 | |||
AK296738.1 | - | 8 | 733 | |||
AK299456.1 | - | 6 | 767 | |||
AK312597.1 | - | 8 | 848 | |||
AL162086.1 | - | 8 | 856 | |||
BC013903.2 | - | 7 | 855 | 2959399 | ||
BC068458.1 | - | 7 | 864 | 4819793 | ||
FJ224318.1 | - | 7 | 840 | |||
J05021.1 | - | 7 | 799 | |||
X51521.1 | - | 7 | 840 |