Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111077.1 | NP_001104547.1 | 7 | 793 | ||
NM_003379.4 | NP_003370.2 | 6 | 827 | |||
XM_011536110.1 | XP_011534412.1 | 3 | 204 | |||
GenBank mRNA | AF187552.1 | - | 1 | 40 | ||
AF188896.1 | - | 1 | 26 | |||
AF189213.1 | - | 1 | 22 | |||
AF190059.1 | - | 1 | 14 | |||
AK296738.1 | - | 7 | 622 | |||
AK299456.1 | - | 5 | 656 | |||
AK312597.1 | - | 7 | 737 | |||
AL162086.1 | - | 7 | 745 | |||
BC013903.2 | - | 6 | 744 | 2959399 | ||
BC068458.1 | - | 6 | 753 | 4819793 | ||
FJ224318.1 | - | 6 | 729 | |||
J05021.1 | - | 6 | 688 | |||
X51521.1 | - | 6 | 729 |