Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003380.3 | NP_003371.2 | 8, 9 | 1690 | ||
XM_006717500.1 | XP_006717563.1 | 7, 8 | 2765 | |||
GenBank mRNA | AF328728.1 | - | 7, 8 | 1406 | ||
AK091813.1 | - | 8, 9 | 1381 | |||
AK097336.1 | - | 8, 9 | 1386 | |||
AK098444.1 | - | 8, 9 | 1260 | |||
AK222482.1 | - | 7, 8 | 1420 | |||
AK222507.1 | - | 7, 8 | 1422 | |||
AK222602.1 | - | 7, 8 | 1422 | |||
AK290643.1 | - | 7, 8 | 1302 | |||
BC000163.2 | - | 7, 8 | 1401 | 2985712 | ||
BC030573.2 | - | 7, 8 | 1401 | 3637336 | ||
BC066956.1 | - | 8, 9 | 1478 | 4823475 | ||
CR407690.1 | - | 7, 8 | 1277 | |||
EU794672.1 | - | 7, 8 | 1422 | |||
KU178388.1 | - | 7, 8 | 1277 | |||
KU178389.1 | - | 8, 9 | 894 | |||
M25246.1 | - | 7, 8 | 781 | |||
X56134.1 | - | 7, 8 | 1320 | |||
Z19554.2 | - | 7, 8 | 1399 |