Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003381.3 | NP_003372.1 | 5 | 564 | ||
NM_194435.2 | NP_919416.1 | 5 | 561 | |||
XM_005267135.3 | XP_005267192.1 | 5 | 513 | |||
XM_006715562.3 | XP_006715625.1 | 5 | 3467 | |||
GenBank mRNA | AK291959.1 | - | 5 | 513 | ||
AK313950.1 | - | 5 | 564 | |||
BC009794.1 | - | 5 | 566 | 4294373 | ||
M36634.1 | - | 5 | 457 | |||
M54930.1 | - | 3 | 244 |