Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pprc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106363.1 | NP_001099833.1 | 5 | 2398 | ||
XM_006231451.2 | XP_006231513.1 | 5 | 2419 | |||
XM_006231452.2 | XP_006231514.1 | 5 | 2418 | |||
XM_006231453.2 | XP_006231515.1 | 5 | 2415 | |||
XM_006231454.2 | XP_006231516.1 | 5 | 2419 | |||
XM_006231455.2 | XP_006231517.1 | 5 | 2410 | |||
XM_006231456.2 | XP_006231518.1 | 5 | 2334 |