Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000638.3 | NP_000629.3 | 8 | 1542 | ||
GenBank mRNA | AK298441.1 | - | 7 | 1591 | ||
AK312968.1 | - | 8 | 1494 | |||
BC005046.1 | - | 8 | 1473 | 4040317 |