Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000638.3 | NP_000629.3 | 3 | 478 | ||
GenBank mRNA | AK298441.1 | - | 3 | 329 | ||
AK312968.1 | - | 3 | 430 | |||
BC005046.1 | - | 3 | 409 | 4040317 | ||
BG470977.1 | - | 3 | 478 | |||
X03168.1 | - | 3 | 390 |