Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009651.1 | NP_001009651.1 | 2 | 341 | ||
GenBank mRNA | BC088182.1 | - | 2 | 341 | 7365286 | |
FQ227497.1 | - | 2 | 293 | |||
FQ231721.1 | - | 2 | 293 | |||
FQ234033.1 | - | 2 | 339 |