Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000268.1 | NP_001000268.1 | 1 | 77 |
Gene Symbol: Olr1675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000894.1 | NP_001000894.1 | 77 |
Gene Symbol: Olr1679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000892.1 | NP_001000892.1 | 77 |