Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000269.1 | NP_001000269.1 | 1 | 241 |
Gene Symbol: Olr1670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000895.1 | NP_001000895.1 | 1 | 241 |
Gene Symbol: Olr1678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000893.1 | NP_001000893.1 | 1 | 241 |
Gene Symbol: Olr1679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000892.1 | NP_001000892.1 | 241 |
Gene Symbol: Olr1680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000891.1 | NP_001000891.1 | 1 | 241 |
Gene Symbol: Olr1682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000890.1 | NP_001000890.1 | 1 | 241 |