Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000271.1 | NP_001000271.1 | 407 |
Gene Symbol: LOC100910894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772701.1 | XP_008770923.1 | 1 | 407 |
Gene Symbol: LOC100910936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772702.1 | XP_008770924.1 | 2 | 463 |
Gene Symbol: Olr1705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000887.1 | NP_001000887.1 | 407 | |||
XM_008772697.1 | XP_008770919.1 | 463 |
Gene Symbol: Olr1708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000276.1 | NP_001000276.1 | 1 | 383 |