Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000274.1 | NP_001000274.1 | 1 | 820 |
Gene Symbol: LOC100910420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751906.2 | XP_003751954.1 | 837 |
Gene Symbol: LOC100910696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772684.1 | XP_008770906.1 | 843 |
Gene Symbol: Olr1689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000889.1 | NP_001000889.1 | 1 | 820 |