Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000275.1 | NP_001000275.1 | 933 |
Gene Symbol: LOC100910382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751905.2 | XP_003751953.1 | 1 | 946 |
Gene Symbol: Olr1691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000273.1 | NP_001000273.1 | 1 | 933 |