Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214456.1 | NP_999621.1 | 1 | 494 |
Gene Symbol: Olr1722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001117.1 | NP_001001117.1 | 1 | 494 |
Gene Symbol: Olr1733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000510.1 | NP_001000510.1 | 1 | 494 |
Gene Symbol: Olr1742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001424.1 | NP_001001424.1 | 1 | 494 |