Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212542.3 | NP_997707.2 | 3 | 545 | ||
XM_006255870.1 | XP_006255932.1 | 3 | 610 | |||
GenBank mRNA | BC079252.1 | - | 3 | 514 | 7133103 | |
EX488885.1 | - | 3 | 539 | |||
FM088354.1 | - | 3 | 544 |