Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212505.2 | NP_997670.1 | 2 | 437 | ||
GenBank mRNA | AW916683.1 | - | 2 | 438 | ||
BC099831.1 | - | 2 | 413 | 7933804 | ||
FM066628.1 | - | 2 | 422 |