Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skiv2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213559.2 | NP_998724.1 | 19 | 2260 | ||
XM_006255923.1 | XP_006255985.1 | 19 | 2323 |