Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098216.1 | NP_001091686.1 | 2, 3 | 441 | ||
GenBank mRNA | BC085938.1 | - | 2, 3 | 394 | ||
BC099077.1 | - | 2, 3 | 441 |