Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694870 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773819.1 | XP_008772041.1 | 8 | 1170 |
Gene Symbol: LOC103690052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772964.1 | XP_008771186.1 | 1170 |
Gene Symbol: Slc5a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106383.1 | NP_001099853.1 | 13 | 1726 |