Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025113.1 | NP_001020284.1 | 13 | 1526 | ||
XM_006256442.2 | XP_006256504.1 | 14 | 1706 | |||
XM_008772879.1 | XP_008771101.1 | 12 | 1381 | |||
GenBank mRNA | AY779029.1 | - | 13 | 1526 |