Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271106.1 | NP_001258035.1 | 6 | 1866 | ||
GenBank mRNA | AJ535460.1 | - | 6 | 1842 |
Set Membership: |
G-protein coupled receptor |