Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182951.1 | NP_891996.1 | 2 | 240 | ||
GenBank mRNA | AY339894.1 | - | 2 | 240 | ||
AY539942.1 | - | 3663 | ||||
BC088852.1 | - | 2 | 251 | 7315523 | ||
DQ103708.1 | - | 2 | 220 |