Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005430.3 | NP_005421.1 | 2 | 334 | ||
GenBank mRNA | BC074798.2 | - | 2 | 172 | 30915522 | |
BC074799.2 | - | 2 | 172 | 30915309 | ||
BT019429.1 | - | 2 | 136 |