Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760641.1 | XP_008758863.1 | 88 | 18689 | ||
GenBank mRNA | FQ219861.1 | - | 2 | 165 |