Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191701.1 | NP_001178630.1 | 2 | 256 | ||
XM_008760662.1 | XP_008758884.1 | 4 | 1048 | |||
XM_008760663.1 | XP_008758885.1 | 4 | 1149 |