Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207595.1 | NP_997478.1 | 6 | 937 | ||
XM_006257186.2 | XP_006257248.1 | 5 | 743 | |||
GenBank mRNA | AY566568.1 | - | 6 | 937 | ||
BC081975.1 | - | 6 | 1126 | 7126874 | ||
FQ231969.1 | - | 6 | 1137 |
Gene Symbol: LOC100910438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760690.1 | XP_008758912.1 | 6 | 1189 | ||
XM_008760691.1 | XP_008758913.1 | 5 | 743 |