Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106406.1 | NP_001099876.1 | 8 | 1509 | ||
XM_006231887.2 | XP_006231949.1 | 8 | 1286 | |||
XM_008760666.1 | XP_008758888.1 | 10 | 1281 | |||
GenBank mRNA | BC078983.1 | - | 8 | 1726 |