Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006522.3 | NP_006513.1 | 4 | 1619 | ||
GenBank mRNA | AB059570.1 | - | 4 | 1409 | ||
AK075522.1 | - | 4 | 1503 | |||
AY009401.1 | - | 4 | 1627 | |||
BC004329.1 | - | 4 | 1407 | 3630498 |
Gene Symbol: WNT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511929.2 | XP_011510231.1 | 1 | 36 |