Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106408.2 | NP_001099878.2 | 11 | 1009 | ||
XM_006231902.2 | XP_006231964.1 | 11 | 1037 | |||
XM_006231903.2 | XP_006231965.1 | 12 | 1220 | |||
XM_006231905.2 | XP_006231967.1 | 10 | 923 | |||
GenBank mRNA | BC158729.1 | - | 11 | 1009 | 9091721 |