Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134436.1 | NP_001127908.1 | 1 | 278 | ||
GenBank mRNA | BC162048.1 | - | 1 | 278 | 7108655 |