Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006976.1 | NP_001006977.1 | 11, 12 | 1387 | ||
XM_006232068.2 | XP_006232130.1 | 12, 13 | 1545 | |||
GenBank mRNA | BC083914.1 | - | 11, 12 | 1387 | 7134406 |