Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053976.1 | NP_446428.1 | 6 | 1136 | ||
GenBank mRNA | BC091275.1 | - | 6 | 1136 | 7383311 |
Gene Symbol: LOC683212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760823.1 | XP_008759045.1 | 2 | 1096 |