Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034129.1 | NP_001029301.1 | 10 | 1256 | ||
XM_006232224.2 | XP_006232286.1 | 6 | 838 | |||
GenBank mRNA | AY325207.1 | - | 10 | 1256 |