Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004626.2 | NP_004617.2 | 4 | 945 | ||
XM_005274231.1 | XP_005274288.1 | 5 | 1201 | |||
XM_011545238.1 | XP_011543540.1 | 5 | 1231 | |||
XM_011545239.2 | XP_011543541.1 | 5 | 1439 | |||
XM_011545240.2 | XP_011543542.1 | 6 | 1395 | |||
XM_011545241.2 | XP_011543543.1 | 4 | 683 | |||
XM_017018241.1 | XP_016873730.1 | 3 | 1703 | |||
XM_017018242.1 | XP_016873731.1 | 3 | 1538 | |||
XM_017018243.1 | XP_016873732.1 | 2 | 357 | |||
GenBank mRNA | AB070218.1 | - | 4 | 884 | ||
AK313570.1 | - | 4 | 947 | |||
BC074790.2 | - | 4 | 864 | 30915564 | ||
BC074791.2 | - | 4 | 864 | 30915347 | ||
BC113386.1 | - | 4 | 865 | 8322438 | ||
BC113388.1 | - | 4 | 865 | 8322440 | ||
BT019492.1 | - | 4 | 822 | |||
JF800675.1 | - | 4 | 546 | |||
JQ715612.1 | - | 4 | 701 | |||
JQ715613.1 | - | 4 | 510 |