Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025407.1 | NP_001020578.1 | 9 | 850 | ||
XM_006232333.1 | XP_006232395.1 | 9 | 1085 | |||
GenBank mRNA | BC098039.1 | - | 9 | 850 | 7456888 |