Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106433.2 | NP_001099903.2 | 4 | 1267 | ||
XM_006232365.2 | XP_006232427.1 | 4 | 1595 | |||
XM_006232366.2 | XP_006232428.1 | 4 | 1586 | |||
XM_006232367.2 | XP_006232429.1 | 4 | 1384 | |||
GenBank mRNA | BC099217.1 | - | 2 | 300 |