Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232841.2 | XP_006232903.1 | 2 | 219 | ||
XM_006232842.2 | XP_006232904.1 | 2 | 219 | |||
XM_006232844.2 | XP_006232906.1 | 2 | 219 | |||
XM_008761252.1 | XP_008759474.1 | 1, 2 | 219 |
Gene Symbol: Pglyrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191961.1 | NP_001178890.1 | 89 | |||
XM_006232834.2 | XP_006232896.1 | 2 | 219 | |||
XM_006232836.2 | XP_006232898.1 | 2 | 219 |