Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106438.1 | NP_001099908.1 | 1 | 76 | ||
XM_006232601.2 | XP_006232663.1 | 2 | 187 |