Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191607.1 | NP_001178536.1 | 3 | 324 | ||
GenBank mRNA | AW141940.1 | - | 3 | 318 |