Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007636.2 | NP_001007637.1 | 1 | 85 | ||
XM_006232603.2 | XP_006232665.1 | 2 | 769 | |||
GenBank mRNA | S68809.1 | - | 1 | 55 |