Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146218.1 | NP_001139690.1 | 2 | 244 | ||
NM_004627.4 | NP_004618.2 | 2 | 354 | |||
GenBank mRNA | AK293113.1 | - | 2 | 309 | ||
AK293866.1 | - | 2 | 287 | |||
AK297186.1 | - | 2 | 314 | |||
AK299144.1 | - | 2 | 197 | |||
AK299245.1 | - | 2 | 315 | |||
AK310268.1 | - | 2 | 245 | |||
BC012415.1 | - | 2 | 287 | 3865141 | ||
BC111393.1 | - | 2 | 243 | |||
DB498697.1 | - | 2 | 245 | |||
DB500750.1 | - | 2 | 354 | |||
Y12478.1 | - | 2 | 298 |
Gene Symbol: WRB-SH3BGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317744.1 | NP_001304673.1 | 2 | 397 | ||
GenBank mRNA | CD514470.1 | - | 2 | 212 | ||
FY213114.1 | - | 2 | 213 | |||
KM576720.1 | - | 2 | 239 |