Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100836.1 | NP_001094306.1 | 5 | 647 | ||
GenBank mRNA | AF416291.1 | - | 2 | 266 | ||
BC158809.1 | - | 5 | 648 | 7364883 | ||
CF113031.1 | - | 2 | 245 | |||
CO555980.1 | - | 5 | 680 |