Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271334.1 | NP_001258263.1 | 2 | 341 | ||
XM_006233003.1 | XP_006233065.1 | 3 | 530 | |||
XM_006233004.2 | XP_006233066.1 | 3 | 504 | |||
XM_006233006.2 | XP_006233068.1 | 4 | 1395 | |||
XM_006233007.2 | XP_006233069.1 | 3 | 1363 | |||
XM_006233008.1 | XP_006233070.1 | 3 | 452 |
Gene Symbol: LOC102550892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233020.2 | XP_006233082.1 | 1191 |