Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024244.1 | NP_001019415.1 | 3 | 528 | ||
GenBank mRNA | BC095842.1 | - | 3 | 528 | 7389454 |