Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079539.1 | NP_001073007.1 | 2 | 359 | ||
NM_005080.3 | NP_005071.2 | 2 | 359 | |||
GenBank mRNA | AB076383.1 | - | 2 | 319 | ||
AB076384.1 | - | 2 | 319 | |||
AK292188.1 | - | 2 | 356 | |||
AK313761.1 | - | 2 | 355 | |||
BC000938.1 | - | 2 | 316 | 3449394 | ||
BC012841.1 | - | 2 | 343 | 3856898 | ||
BC015709.1 | - | 2 | 252 | |||
BI545390.1 | - | 2 | 380 | |||
CR456611.1 | - | 2 | 314 | |||
M31627.1 | - | 2 | 320 |