Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191569.1 | NP_001178498.1 | 11 | 2041 | ||
XM_006233069.1 | XP_006233131.1 | 11 | 2032 | |||
XM_006233070.2 | XP_006233132.1 | 11 | 2359 | |||
XM_006233071.2 | XP_006233133.1 | 11 | 2144 |