Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106467.3 | NP_001099937.1 | 570 | |||
GenBank mRNA | BC160912.1 | - | 559 | 7456962 |
Gene Symbol: NEWGENE_1305281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233223.2 | XP_006233285.1 | 5 | 702 | ||
XM_006233224.2 | XP_006233286.1 | 5 | 817 | |||
XM_008761473.1 | XP_008759695.1 | 5 | 573 |