Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102556447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233345.2 | XP_006233407.1 | 4 | 780 | ||
XM_008761533.1 | XP_008759755.1 | 4 | 780 |
Gene Symbol: Arhgef38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761544.1 | XP_008759766.1 | 428 |