Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 9, 10 | 1976 | ||
NR_027299.1 | 8, 9 | 1852 | ||||
GenBank mRNA | AK222844.1 | - | 8, 9 | 1669 | ||
AK289761.1 | - | 9, 10 | 1905 | |||
AK295711.1 | - | 9, 10 | 1794 | |||
AK311039.1 | - | 8, 9 | 1782 | |||
BC016620.1 | - | 9, 10 | 1887 | 4509957 | ||
D21089.1 | - | 9, 10 | 1977 | |||
EU530523.1 | - | 1, 2 | 429 | |||
EU530532.1 | - | 1, 2 | 332 | |||
HM113488.1 | - | 9, 10 | 1876 | |||
KJ534962.1 | - | 9, 10 | 1872 | |||
X65024.1 | - | 8, 9 | 1712 |