Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044232.1 | NP_001037697.1 | 6 | 642 | ||
XM_006233368.2 | XP_006233430.1 | 6 | 2316 | |||
XM_006233369.2 | XP_006233431.1 | 6 | 862 | |||
GenBank mRNA | BC099190.1 | - | 6 | 642 | 7383412 | |
FQ232089.1 | - | 6 | 658 |